missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KAL1 Partial ORF (NP_000207, 548 a.a. - 657 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000207 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3730 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137071
|
Abnova™
H00003730-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16127071
|
Abnova™
H00003730-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Mutations in this gene cause the X-linked Kallmann syndrome. The encoded protein is similar in sequence to proteins known to function in neural cell adhesion and axonal migration. In addition, this cell surface protein is N-glycosylated and may have anti-protease activity. [provided by RefSeq]
Sequence: LAKPENLSASFIVQDVNITGHFSWKMAKANLYQPMTGFQVTWAEVTTESRQNSLPNSIISQSQILPSDHYVLTVPNLRPSTLYRLEVQVLTPGGEGPATIKTFRTPELPPSpecifications
NP_000207 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADMLX/HHA/KAL/KALIG-1/KMS | |
KAL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3730 | |
KAL1 (Human) Recombinant Protein (Q01) | |
LAKPENLSASFIVQDVNITGHFSWKMAKANLYQPMTGFQVTWAEVTTESRQNSLPNSIISQSQILPSDHYVLTVPNLRPSTLYRLEVQVLTPGGEGPATIKTFRTPELPP | |
RUO | |
KAL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |