missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITGB3BP Partial ORF (NP_055103, 78 a.a. - 177 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023421-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16025335

  • 335.00€ / 10µg
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Sequence: STTKDNDEFMMLLSKVEKLSEEIMEIMQNLSSIQALEGSRELENLIGISCASHFLKREMQKTKELMTKVNKQKLFEKSTGLPHKASRHLDSYEFLKAILN
Specifications

Specifications

NP_055103
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
STTKDNDEFMMLLSKVEKLSEEIMEIMQNLSSIQALEGSRELENLIGISCASHFLKREMQKTKELMTKVNKQKLFEKSTGLPHKASRHLDSYEFLKAILN
RUO
ITGB3BP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23421
ITGB3BP (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CENP-R/CENPR/HSU37139/NRIF3/TAP20
ITGB3BP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers