Learn More
Abnova™ Human IRX2 Full-length ORF (NP_150366.1, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00153572-P01.10ug
Additional Details : Weight : 0.00010kg
Description
IRX2 is a member of the Iroquois homeobox gene family. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.[supplied by OMIM]
Sequence: MSYPQGYLYQAPGSLALYSCPAYGASALAAPRSEELARSASGSAFSPYPGSAAFTAQAATGFGSPLQYSADAAAAAAGFPSYMGAPYDAHTTGMTGAISYHPYGSAAYPYQLNDPAYRKNATRDATATLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRNKSEDEDEDEGDATRSKDESPDKAQEGTETSAEDEGISLHVDSLTDHSCSAESDGEKLPCRAGDPLCESGSECKDKYDDLEDDEDDDEEGERGLAPPKPVTSSPLTGLEAPLLSPPPEAAPRGGRKTPQGSRTSPGAPPPASKPKLWSLAEIATSDLKQPSLGPGCGPPGLPAAAAPASTGAPPGGSPYPASPLLGRPLYYTSPFYGNYTNYGNLNAALQGQGLLRYNSAAAAPGEALHTAPKAASDAGKAGAHPLESHYRSPGGGYEPKKDASEGCTVVGGGVQPYLSpecifications
NP_150366.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IRXA2 | |
IRX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
153572 | |
IRX2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSYPQGYLYQAPGSLALYSCPAYGASALAAPRSEELARSASGSAFSPYPGSAAFTAQAATGFGSPLQYSADAAAAAAGFPSYMGAPYDAHTTGMTGAISYHPYGSAAYPYQLNDPAYRKNATRDATATLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRNKSEDEDEDEGDATRSKDESPDKAQEGTETSAEDEGISLHVDSLTDHSCSAESDGEKLPCRAGDPLCESGSECKDKYDDLEDDEDDDEEGERGLAPPKPVTSSPLTGLEAPLLSPPPEAAPRGGRKTPQGSRTSPGAPPPASKPKLWSLAEIATSDLKQPSLGPGCGPPGLPAAAAPASTGAPPGGSPYPASPLLGRPLYYTSPFYGNYTNYGNLNAALQGQGLLRYNSAAAAPGEALHTAPKAASDAGKAGAHPLESHYRSPGGGYEPKKDASEGCTVVGGGVQPYL | |
RUO | |
IRX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |