Learn More
ACROBiosystems Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein, His Tag&Tag Free
Brand:  ACROBiosystems IT1-H52W7-100UG
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15817147
  • 413.60€ / 100µg
Estimated Shipment: 19-10-2021
Add to basket
Description
Description
Integrin alpha 6 beta 1, also called platelet glycoprotein GPIc-IIa, is a receptor for laminin on platelets. The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. ITGA6 has been shown to interact with TSPAN4 and GIPC1.
Human ITGA6&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA6 and ITGB1, has a calculated MW of 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1). Subunit ITGA6 is fused with an acidic tail at the C-terminus and followed by a polyhistid
Specifications
Specifications
NP_000201.2 (ITGA6) and NP_002202.2 (ITGB1)
HEK293
Human
116.8 kDa (ITGA6) and 83.7 kDa (ITGB1)
100 µg
Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1)
Integrin alpha 6 beta 1,ITGA6andITGB1
Unconjugated
50 mM Tris, 150 mM NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>90% as determined by SDS-PAGE
Human ITGA6&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1) (Accession # P23229-2 (ITGA6) and P05556-1 (ITGB1)).
-20°C
Integrin alpha 6 beta 1
His Tag