Learn More
Abnova™ Human ING3 Full-length ORF (NP_061944.2, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_061944.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54556 |
Molecular Weight (g/mol) | 73.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16110653
|
Abnova™
H00054556-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16120653
|
Abnova™
H00054556-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed. [provided by RefSeq]
Sequence: MLYLEDYLEMIEQLPMDLRDRFTEMREMDLQVQNAMDQLEQRVSEFFMNAKKNKPEWREEQMASIKKDYYKALEDADEKVQLANQIYDLVDRHLRKLDQELAKFKMELEADNAGITEILERRSLELDTPSQPVNNHHAHSHTPVEKRKYNPTSHHTTTDHIPEKKFKSEALLSTLTSDASKENTLGCRNNNSTASSNNAYNVNSSQPLGSYNIGSLSSGTGAGAITMAAAQAVQATAQMKEGRRTSSLKASYEAFKNNDFQLGKEFSMARETVGYSSSSALMTTLTQNASSSAADSRSGRKSKNNNKSSSQQSSSSSSSSSLSSCSSSSTVVQEISQQTTVVPESDSNSQVDWTYDPNEPRYCICNQVSYGEMVGCDNQDCPIEWFHYGCVGLTEAPKGKWYCPQCTAAMKRRGSRHKSpecifications
NP_061944.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLYLEDYLEMIEQLPMDLRDRFTEMREMDLQVQNAMDQLEQRVSEFFMNAKKNKPEWREEQMASIKKDYYKALEDADEKVQLANQIYDLVDRHLRKLDQELAKFKMELEADNAGITEILERRSLELDTPSQPVNNHHAHSHTPVEKRKYNPTSHHTTTDHIPEKKFKSEALLSTLTSDASKENTLGCRNNNSTASSNNAYNVNSSQPLGSYNIGSLSSGTGAGAITMAAAQAVQATAQMKEGRRTSSLKASYEAFKNNDFQLGKEFSMARETVGYSSSSALMTTLTQNASSSAADSRSGRKSKNNNKSSSQQSSSSSSSSSLSSCSSSSTVVQEISQQTTVVPESDSNSQVDWTYDPNEPRYCICNQVSYGEMVGCDNQDCPIEWFHYGCVGLTEAPKGKWYCPQCTAAMKRRGSRHK | |
RUO | |
ING3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54556 | |
ING3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Eaf4/FLJ20089/ING2/p47ING3 | |
ING3 | |
Recombinant | |
wheat germ expression system |