missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IL18RAP Partial ORF (NP_003844, 20 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_003844
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8807
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16149315
View Documents Promotion Details
Abnova™
H00008807-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16159315
View Documents Promotion Details
Abnova™
H00008807-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

The protein encoded by this gene is an accessory subunit of the heterodimeric receptor for IL18. This protein enhances the IL18 binding activity of IL18R1 (IL1RRP), a ligand binding subunit of IL18 receptor. The coexpression of IL18R1 and this protein is required for the activation of NF-kappaB and MAPK8 (JNK) in response to IL18. [provided by RefSeq]

Sequence: FNISGCSTKKLLWTYSTRSEEEFVLFCDLPEPQKSHFCHRNRLSPKQVPEHLPFMGSNDLSDVQWYQQPSNGDPLEDIRKSYPHIIQDKCTLHFLTPGVNNSGSYICRPK
Specifications

Specifications

NP_003844
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACPL/CD218b/CDw218b/IL18RB/MGC120589/MGC120590
IL18RAP
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8807
IL18RAP (Human) Recombinant Protein (Q01)
FNISGCSTKKLLWTYSTRSEEEFVLFCDLPEPQKSHFCHRNRLSPKQVPEHLPFMGSNDLSDVQWYQQPSNGDPLEDIRKSYPHIIQDKCTLHFLTPGVNNSGSYICRPK
RUO
IL18RAP
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications