missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IL-21 R Protein, Fc Tag
404.99€ - 1593.84€
Specifications
Accession Number | AAH04348 |
---|---|
Molecular Weight (g/mol) | 51.6 kDa |
Source | Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348). |
Immunogen | Cys 20 - Pro 236 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15817127
|
ACROBiosystems
ILR-H5257-200UG |
200 μg |
404.99€
200µg |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
15807127
|
ACROBiosystems
ILR-H5257-1MG |
1 mg |
1593.84€
1mg |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
Description
Interleukin-21 receptor (IL21R) is also known as CD antigen CD360, Novel interleukin receptor (NILR), is a receptor for interleukin-21 and belongs to the type I cytokine receptor family or type 4 subfamily. IL21R contains two fibronectin type-III domains. IL21R is selectively expressed in lymphoid tissues and most highly expressed in thymus and spleen. IL21R transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 120-130 kDa under non-reducing (NR) condition (SDS-PSpecifications
AAH04348 | |
Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348). | |
-20°C | |
IL-21 R | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
51.6 kDa | |
Cys 20 - Pro 236 | |
IL21R,CD360,NILR | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |