missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-21 R Protein, Fc Tag

Brand:  ACROBiosystems ILR-H5257-200UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817127

  • 312.72€ / 200µg


Estimated Shipment: 20-10-2021
Add to basket

Description

Description

Interleukin-21 receptor (IL21R) is also known as CD antigen CD360, Novel interleukin receptor (NILR), is a receptor for interleukin-21 and belongs to the type I cytokine receptor family or type 4 subfamily. IL21R contains two fibronectin type-III domains. IL21R is selectively expressed in lymphoid tissues and most highly expressed in thymus and spleen. IL21R transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 120-130 kDa under non-reducing (NR) condition (SDS-P
Specifications

Specifications

AAH04348
HEK293
Human
51.6 kDa
200 µg
Cys 20 - Pro 236
IL21R,CD360,NILR
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>92% as determined by SDS-PAGE
Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).
-20°C
IL-21 R
Fc Tag
Videos
SDS
Documents
Certificates