missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IgG1 Fc Protein, His Tag

365.80€ - 1214.97€

Specifications

Molecular Weight (g/mol) 26.9 kDa
Source Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1).
Immunogen Glu 99 - Lys 330
Storage Requirements -20°C
Gene Alias IgG1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15816987
View Documents Promotion Details
ACROBiosystems
IG1-H5225-200UG
200 μg
365.80€
200µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
15806987
View Documents Promotion Details
ACROBiosystems
IG1-H5225-1MG
1 mg
1214.97€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
Description

Description

Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

26.9 kDa
Glu 99 - Lys 330
IgG1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1).
-20°C
IgG1 Fc
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Other Recombinant Proteins
Videos
SDS
Documents

Documents

Product Certifications