Learn More
ACROBiosystems Human IgG1 Fc Protein, His Tag
Brand: ACROBiosystems IG1-H5225-200UG
Additional Details : Weight : 0.00001kg
Description
Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
26.9 kDa | |
Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1). | |
-20°C | |
IgG1 Fc | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
200 μg | |
Glu 99 - Lys 330 | |
IgG1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |