Learn More
ACROBiosystems Human IFN-alpha/beta R2 Protein, His Tag
Brand: ACROBiosystems IF2-H5224-1MG
Additional Details : Weight : 0.00001kg
Description
Interferon alpha/beta receptor 2 (IFNAR2) is also known as IFN-alpha binding protein, IFN-alpha/beta receptor 2, Type I interferon receptor 2, IFNABR and IFNARB, which is a single-pass type I membrane protein and belongs to the type II cytokine receptor family. IFNAR2 can associate with IFNAR1 to form the type I interferon receptor. IFNAR2 is a receptor for interferons alpha and beta.IFNAR2 involves in IFN-mediated STAT1, STAT2 and STAT3 activation. Isoform 1 and isoform 2 of IFNAR2 are directly involved in signal transduction due to their association with the TYR kinase, JAK1. Isoform 3 of IFNAR2 is a potent inhibitor of type I IFN receptor activity.Genetic variations in IFNAR2 influence susceptibility to hepatitis B virus (HBV) infection.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 34-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_997467.1 | |
1 mg | |
Ile 27 - Lys 243 | |
IFNAR2,IFNARB,IFNABR,IFN-R-2,IFN-alpha/beta receptor 2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
25.8 kDa | |
Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # P48551-2). | |
-20°C | |
IFN-alpha/beta R2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |