missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IER5 Partial ORF (NP_057629, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_057629
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51278
Molecular Weight (g/mol) 31.9kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16116786
View Documents Promotion Details
Abnova™
H00051278-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16106786
View Documents Promotion Details
Abnova™
H00051278-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation. [provided by RefSeq]

Sequence: MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSDPCPGLY
Specifications

Specifications

NP_057629
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.9kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC102760/SBBI48
IER5
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
51278
IER5 (Human) Recombinant Protein (Q01)
MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSDPCPGLY
RUO
IER5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications