Learn More
Abnova™ Human HOXA5 Partial ORF (NP_061975.1, 171 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_061975.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3202 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024155
|
Abnova™
H00003202-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16014155
|
Abnova™
H00003202-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis. [provided by RefSeq]
Sequence: PAQPQIYPWMRKLHISHDNIGGPEGKRARTAYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLSERQIKIWFQNRRMKWKKDNKLKSMSMAAAGGAFRPSpecifications
NP_061975.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX1/HOX1.3/HOX1C/MGC9376 | |
HOXA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3202 | |
HOXA5 (Human) Recombinant Protein (Q01) | |
PAQPQIYPWMRKLHISHDNIGGPEGKRARTAYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLSERQIKIWFQNRRMKWKKDNKLKSMSMAAAGGAFRP | |
RUO | |
HOXA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |