missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HOMER2 Full-length ORF (NP_004830.2, 1 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004830.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9455 |
Molecular Weight (g/mol) | 65.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173682
|
Abnova™
H00009455-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16163682
|
Abnova™
H00009455-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein may be involved in cell growth. Four transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MGEQPIFTTRAHVFQIDPNTKKNWMPASKQAVTVSYFYDVTRNSYRIISVDGAKVIINSTITPNMTFTKTSQKFGQWADSRANTVFGLGFSSEQQLTKFAEKFQEVKEAAKIAKDKTQEKIETSSNHSQASSVNGTDDEKASHAGPANTHLKSENDKLKIALTQSAANVKKWEIELQTLRESNARLTTALQESAASVEQWKRQFSICRDENDRLRNKIDELEEQCSEINREKEKNTQLKRRIEELEAELREKETELKDLRKQSEIIPQLMSECEYVSEKLEAAERDNQNLEDKVRSLKTDIEESKYRQRHLKVELKSFLEVLDGKIDDLHDFRRGLSKLGTDNSpecifications
NP_004830.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGEQPIFTTRAHVFQIDPNTKKNWMPASKQAVTVSYFYDVTRNSYRIISVDGAKVIINSTITPNMTFTKTSQKFGQWADSRANTVFGLGFSSEQQLTKFAEKFQEVKEAAKIAKDKTQEKIETSSNHSQASSVNGTDDEKASHAGPANTHLKSENDKLKIALTQSAANVKKWEIELQTLRESNARLTTALQESAASVEQWKRQFSICRDENDRLRNKIDELEEQCSEINREKEKNTQLKRRIEELEAELREKETELKDLRKQSEIIPQLMSECEYVSEKLEAAERDNQNLEDKVRSLKTDIEESKYRQRHLKVELKSFLEVLDGKIDDLHDFRRGLSKLGTDN | |
RUO | |
HOMER2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9455 | |
HOMER2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACPD/CPD/HOMER-2/HOMER2A/HOMER2B/Vesl-2 | |
HOMER2 | |
Recombinant | |
wheat germ expression system |