missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HIPK3 Partial ORF (NP_005725, 1018 a.a. - 1117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010114-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16025225

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Sequence: CQPLIKGRSAPGRLNQPSAVGTRQQKLTSAFQQQHLNFSQVQHFGSGHQEWNGNFGHRRQQAYIPTSVTSNPFTLSHGSPNHTAVHAHLAGNTHLGGQPT
Specifications

Specifications

NP_005725
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CQPLIKGRSAPGRLNQPSAVGTRQQKLTSAFQQQHLNFSQVQHFGSGHQEWNGNFGHRRQQAYIPTSVTSNPFTLSHGSPNHTAVHAHLAGNTHLGGQPT
RUO
HIPK3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10114
HIPK3 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DYRK6/FIST3/PKY/YAK1
HIPK3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers