Learn More
Abnova™ Human GRB14 Partial ORF (NP_004481, 25 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004481 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2888 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16195731
|
Abnova™
H00002888-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16105741
|
Abnova™
H00002888-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date. [provided by RefSeq]
Sequence: QVCGAAQGRGDAHDLAPAPWLHARALLPLPDGTRGCAADRRKKKDLDVPEMPSIPNPFPELCCSPITSVLSADLFPKANSRKKQVIKVYSEDETSRALDVSpecifications
NP_004481 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRB14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2888 | |
GRB14 (Human) Recombinant Protein (Q01) | |
QVCGAAQGRGDAHDLAPAPWLHARALLPLPDGTRGCAADRRKKKDLDVPEMPSIPNPFPELCCSPITSVLSADLFPKANSRKKQVIKVYSEDETSRALDV | |
RUO | |
GRB14 | |
Recombinant | |
wheat germ expression system |