missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human GPA33 / A33 Protein, His Tag

Brand:  ACROBiosystems GP3-H5224-1MG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15816877

  • 1714.94€ / 1mg


Estimated Shipment: 20-10-2021
Add to basket

Description

Description

Glycoprotein A33 (GPA33) is also known as Cell surface A33 antigen, is a single-pass type I membrane protein which is expressed in normal gastrointestinal epithelium and in 95% of colon cancers. GPA33 The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily, which contains. GPA33 may play a role in cell-cell recognition and signaling.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

AAH74830
HEK293
Human
24.4 kDa
1 mg
Ile 22 - Val 235
GPA33,A33
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>96% as determined by SDS-PAGE
Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830).
-20°C
GPA33
His Tag
Videos
SDS
Documents
Certificates