missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FRMD3 Partial ORF (NP_777598.2, 393 a.a. - 480 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00257019-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192627

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Protein 4.1 of red blood cells, or 4.1R (EPB41; MIM 130500), is a multifunctional protein essential for maintaining erythrocyte shape and membrane mechanical properties. The protein 4.1 family comprises a group of structural proteins that includes, in addition to 4.1R, 4.1G (general type; MIM 603237), 4.1B (brain type; MIM 605331), 4.1N (neuron type; MIM 602879), and 4.1O (ovary type).[supplied by OMIM]

Sequence: SAPLISSSPVKAAREYEDPPSEEEDKIKEEPLTISELVYNPSASLLPTPVDDDEIDMLFDCPSRLELEREDTDSFEDLEADENAFLIA
Specifications

Specifications

NP_777598.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SAPLISSSPVKAAREYEDPPSEEEDKIKEEPLTISELVYNPSASLLPTPVDDDEIDMLFDCPSRLELEREDTDSFEDLEADENAFLIA
RUO
FRMD3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
257019
FRMD3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4.1O/EPB41L4O/EPB41LO/MGC20553/P410
FRMD3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers