Learn More
Abnova™ Human FHOD1 Partial ORF (NP_037373.1, 2 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_037373.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29109 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116026
|
Abnova™
H00029109-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16106026
|
Abnova™
H00029109-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein which is a member of the formin/diaphanous family of proteins. The gene is ubiquitously expressed but is found in abundance in the spleen. The encoded protein has sequence homology to diaphanous and formin proteins within the Formin Homology (FH)1 and FH2 domains. It also contains a coiled-coil domain, a collagen-like domain, two nuclear localization signals, and several potential PKC and PKA phosphorylation sites. It is a predominantly cytoplasmic protein and is expressed in a variety of human cell lines. [provided by RefSeq]
Sequence: AGGEDRGDGEPVSVVTVRVQYLEDTDPFACANFPEPRRAPTCSLDGALPLGAQIPAVHRLLGAPLKLEDCALQVSPSGYYLDTELSLEEQREMLEGFYSpecifications
NP_037373.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FHOS | |
FHOD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
29109 | |
FHOD1 (Human) Recombinant Protein (Q01) | |
AGGEDRGDGEPVSVVTVRVQYLEDTDPFACANFPEPRRAPTCSLDGALPLGAQIPAVHRLLGAPLKLEDCALQVSPSGYYLDTELSLEEQREMLEGFY | |
RUO | |
FHOD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |