Learn More
ACROBiosystems Human FGF acidic Protein, Tag Free
Brand: ACROBiosystems AFF-H4116-1MG
Additional Details : Weight : 0.00001kg
Description
Heparin-binding growth factor 1 is a protein that in humans is encoded by the FGF1 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF acidic is a potent growth factor for fibroblasts and endothelial cells. FGF acidic is involved in wound repair, angiogenesis, and development. FGF acidic is secreted from cells via an endoplasmic reticulum/Golgi independent mechanism. The ability of FGF acidic to bind to heparin sulfate is required for its ability to interact with FGF receptors and induce signaling. There are four distinct FGF receptors and each has multiple splice variants. FGF acidic binds with high affinity to many, but not all, FGFRs. Signaling cascades activated through FGF basic binding to FGFR include the ras-raf-MAPK, PLCγ/PKC, and PI3K/Akt pathways.
This protein carries no "e;tag"e;. The protein has a calculated MW of 16.0 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).Specifications
NP_000791.1 | |
1 mg | |
Phe 16 - Asp 155 | |
FGF1,AFGF,ECGF,ECGF-beta,ECGFA,ECGFB,FGF-alpha,FGFA,GLIO703,HBGF1 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |
16.0 kDa | |
Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession # P05230-1). | |
-20°C | |
FGF acidic | |
Human | |
PBS, pH7.4 | |
E.coli | |
Cytokines and Growth Factors |