missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FBXO36 Partial ORF (NP_777559, 66 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_777559 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 130888 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101797
|
Abnova™
H00130888-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16191787
|
Abnova™
H00130888-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: HLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNSpecifications
NP_777559 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37592/FLJ41090/Fbx36 | |
FBXO36 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
130888 | |
FBXO36 (Human) Recombinant Protein (Q01) | |
HLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTN | |
RUO | |
FBXO36 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |