Learn More
Abnova™ Human FANCC Full-length ORF (NP_000127.2, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000127.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2176 |
Molecular Weight (g/mol) | 89.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107667
|
Abnova™
H00002176-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16117667
|
Abnova™
H00002176-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group C. [provided by RefSeq]
Sequence: MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKEPQNSGQSKLNSWIQGVLSHILSALRFDKEVALFTQGLGYAPIDYYPGLLKNMVLSLASELRENHLNGFNTQRRMAPERVASLSRVCVPLITLTDVDPLVEALLICHGREPQEILQPEFFEAVNEAILLKKISLPMSAVVCLWLRHLPSLEKAMLHLFEKLISSERNCLRRIECFIKDSSLPQAACHPAIFRVVDEMFRCALLETDGALEIIATIQVFTQCFVEALEKASKQLRFALKTYFPYTSPSLAMVLLQDPQDIPRGHWLQTLKHISELLREAVEDQTHGSCGGPFESWFLFIHFGGWAEMVAEQLLMSAAEPPTALLWLLAFYYGPRDGRQQRAQTMVQVKAVLGHLLAMSRSSSLSAQDLQTVAGQGTDTDLRAPAQQLIRHLLLNFLLWAPGGHTIAWDVITLMAHTAEITHEIIGFLDQTLYRWNRLGIESPRSEKLARELLKELRTQVSpecifications
NP_000127.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKEPQNSGQSKLNSWIQGVLSHILSALRFDKEVALFTQGLGYAPIDYYPGLLKNMVLSLASELRENHLNGFNTQRRMAPERVASLSRVCVPLITLTDVDPLVEALLICHGREPQEILQPEFFEAVNEAILLKKISLPMSAVVCLWLRHLPSLEKAMLHLFEKLISSERNCLRRIECFIKDSSLPQAACHPAIFRVVDEMFRCALLETDGALEIIATIQVFTQCFVEALEKASKQLRFALKTYFPYTSPSLAMVLLQDPQDIPRGHWLQTLKHISELLREAVEDQTHGSCGGPFESWFLFIHFGGWAEMVAEQLLMSAAEPPTALLWLLAFYYGPRDGRQQRAQTMVQVKAVLGHLLAMSRSSSLSAQDLQTVAGQGTDTDLRAPAQQLIRHLLLNFLLWAPGGHTIAWDVITLMAHTAEITHEIIGFLDQTLYRWNRLGIESPRSEKLARELLKELRTQV | |
RUO | |
FANCC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2176 | |
FANCC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FA3/FAC/FACC/FLJ14675 | |
FANCC | |
Recombinant | |
wheat germ expression system |