Learn More
ACROBiosystems Human FABP3 / H-FABP Protein, His Tag
323.90€ - 1567.72€
Specifications
Accession Number | NP_004093 |
---|---|
Molecular Weight (g/mol) | 15.7 kDa |
Source | Human FABP3, His Tag (FA3-H5128) is expressed from E.coli cells. It contains AA Met 1 - Ala 133 (Accession # NP_004093). |
Immunogen | Met 1 - Ala 133 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15826697
|
ACROBiosystems
FA3-H5128-100UG |
100 μg |
323.90€
100µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
15836697
|
ACROBiosystems
FA3-H5128-1MG |
1 mg |
1567.72€
1mg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
Description
Fatty acid-binding protein 3 (FABP3) is also known as Heart-type fatty acid binding protein (H-FABP), Mammary-derived growth inhibitor (MDGI), Muscle fatty acid-binding protein (M-FABP), FABP11, which belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. FABP are thought to play a role in the intracellular transport of long-chain fatty acids and their acyl-CoA esters. H-FABP / FABP3 is involved in active fatty acid metabolism where it transports fatty acids from the cell membrane to mitochondria for oxidation. FABP3 may also contribute to AS160 phosphorylation by maintaining insulin-dependent Akt activation in the cells under a lipotoxic condition.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 15-17 kDa under reducing (R) condition (SDS-PAGE).Specifications
NP_004093 | |
Human FABP3, His Tag (FA3-H5128) is expressed from E.coli cells. It contains AA Met 1 - Ala 133 (Accession # NP_004093). | |
-20°C | |
FABP3 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
E.coli | |
Bio-Markers and CD Antigens |
15.7 kDa | |
Met 1 - Ala 133 | |
FABP3,H-FABP,FABP11,MDGI,M-FABP,hFABP,O-FABP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |