Learn More
Abnova™ Human EPHA4 Full-length ORF (no Protein_acc, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002043-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]
Sequence: MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSPSpecifications
no protein_acc | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.18kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HEK8/SEK/TYRO1 | |
EPHA4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2043 | |
EPHA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP | |
RUO | |
EPHA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |