missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EIF3M Full-length ORF (NP_006351.2, 1 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_006351.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10480 |
Molecular Weight (g/mol) | 68.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105182
|
Abnova™
H00010480-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16195172
|
Abnova™
H00010480-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
HFLB5 encodes a broadly expressed protein containing putative membrane fusion domains that acts as a receptor or coreceptor for entry of herpes simplex virus (HSV) (Perez et al., 2005 [PubMed 15919898]).[supplied by OMIM]
Sequence: MSVPAFIDISEEDQAAELRAYLKSKGAEISEENSEGGLHVDLAQIIEACDVCLKEDDKDVESVMNSVVSLLLILEPDKQEALIESLCEKLVKFREGERPSLRLQLLSNLFHGMDKNTPVRYTVYCSLIKVAASCGAIQYIPTELDQVRKWISDWNLTTEKKHTLLRLLYEALVDCKKSDAASKVMVELLGSYTEDNASQARVDAHRCIVRALKDPNAFLFDHLLTLKPVKFLEGELIHDLLTIFVSAKLASYVKFYQNNKDFIDSLGLLHEQNMAKMRLLTFMGMAVENKEISFDTMQQELQIGADDVEAFVIDAVRTKMVYCKIDQTQRKVVVSHSTHRTFGKQQWQQLYDTLNAWKQNLNKVKNSLLSLSDTSpecifications
NP_006351.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVPAFIDISEEDQAAELRAYLKSKGAEISEENSEGGLHVDLAQIIEACDVCLKEDDKDVESVMNSVVSLLLILEPDKQEALIESLCEKLVKFREGERPSLRLQLLSNLFHGMDKNTPVRYTVYCSLIKVAASCGAIQYIPTELDQVRKWISDWNLTTEKKHTLLRLLYEALVDCKKSDAASKVMVELLGSYTEDNASQARVDAHRCIVRALKDPNAFLFDHLLTLKPVKFLEGELIHDLLTIFVSAKLASYVKFYQNNKDFIDSLGLLHEQNMAKMRLLTFMGMAVENKEISFDTMQQELQIGADDVEAFVIDAVRTKMVYCKIDQTQRKVVVSHSTHRTFGKQQWQQLYDTLNAWKQNLNKVKNSLLSLSDT | |
RUO | |
EIF3M | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10480 | |
EIF3M (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B5/FLJ29030/GA17/PCID1/hfl-B5 | |
EIF3M | |
Recombinant | |
wheat germ expression system |