Learn More
Abnova™ Human ECT2 Partial ORF (AAH06838.1, 46 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH06838.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1894 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013925
|
Abnova™
H00001894-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16003925
|
Abnova™
H00001894-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis. [provided by RefSeq]
Sequence: LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSRSpecifications
AAH06838.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10461/MGC138291 | |
ECT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1894 | |
ECT2 (Human) Recombinant Protein (Q01) | |
LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSR | |
RUO | |
ECT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |