missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DSE Partial ORF (NP_037484, 574 a.a. - 673 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_037484 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29940 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126166
|
Abnova™
H00029940-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16116166
|
Abnova™
H00029940-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is localized to the endoplasmic reticulum. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: LGEESPLETAASFFHNVDVPFEETVVDGVHGAFIRQRDGLYKMYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQSSpecifications
NP_037484 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DSEPI/SART2 | |
DSE | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
29940 | |
DSE (Human) Recombinant Protein (Q01) | |
LGEESPLETAASFFHNVDVPFEETVVDGVHGAFIRQRDGLYKMYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQS | |
RUO | |
DSE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |