missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOK2 Full-length ORF (AAH32623, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009046-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193212

  • 349.00€ / 10µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia (CML) patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. This encoded protein binds p120 (RasGAP) from CML cells. [provided by RefSeq]

Sequence: MGDGAVKQGFLYLQQQQTFGKKWRRFGASLYGGSDCALARLELQEGPEKPRRCGAARKVIRLSDCLRVAEAGGEASSPRDTSAFFLETKERLYLLAAPAAERGDWVQAICLLAFPGQRKELSGPEGKQSRPCMEENELYSSAVTVGPHKEFAVTMRPTEASERCHLRGSYTLRAGESALELWGGPEPGTQLYDWPYRFLRRFGRDKVTFSFEAGRRCVSGEGNFEFETRQGNEIFLALEEAISAQKNAAPATPQPQPATIPASLPRPDSPYSRPHDSLPPPSPTTPVPAPRPRGQEGEYAVPFDAVARSLGKNFRGILAVPPQLLADPLYDSIEETLPPRPDHIYDEPEGVAALSLYDSPQEPRGEAWRRQATADRDPAGLQHVQPAGQDFSASGWQPGTEYDNVVLKKGPK
Specifications

Specifications

AAH32623
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.06kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
p56DOK/p56dok-2
DOK2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9046
DOK2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGDGAVKQGFLYLQQQQTFGKKWRRFGASLYGGSDCALARLELQEGPEKPRRCGAARKVIRLSDCLRVAEAGGEASSPRDTSAFFLETKERLYLLAAPAAERGDWVQAICLLAFPGQRKELSGPEGKQSRPCMEENELYSSAVTVGPHKEFAVTMRPTEASERCHLRGSYTLRAGESALELWGGPEPGTQLYDWPYRFLRRFGRDKVTFSFEAGRRCVSGEGNFEFETRQGNEIFLALEEAISAQKNAAPATPQPQPATIPASLPRPDSPYSRPHDSLPPPSPTTPVPAPRPRGQEGEYAVPFDAVARSLGKNFRGILAVPPQLLADPLYDSIEETLPPRPDHIYDEPEGVAALSLYDSPQEPRGEAWRRQATADRDPAGLQHVQPAGQDFSASGWQPGTEYDNVVLKKGPK
RUO
DOK2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers