missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DOC2B Full-length ORF (AAI53025.1, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAI53025.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8447 |
Molecular Weight (g/mol) | 72.27kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162462
|
Abnova™
H00008447-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16152462
|
Abnova™
H00008447-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca(2+)-dependent intracellular vesicle trafficking in various types of cells. [provided by RefSeq]
Sequence: MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSDSpecifications
AAI53025.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.27kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSD | |
RUO | |
DOC2B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8447 | |
DOC2B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DOC2B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |