missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DO Partial ORF (NP_066549, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_066549 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 420 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16104924
|
Abnova™
H00000420-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16114924
|
Abnova™
H00000420-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that contains a mono-ADP-ribosylation (ART) motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known. [provided by RefSeq]
Sequence: SFDDQYQGCSKQVMEKLTQGDYFTKDIEAQKNYFRMWQKAHLAWLNQGKVLPQNMTTTHAVAILFYTLNSNVHSDFTRAMASVARTPQQYERSFHFKYLHSpecifications
NP_066549 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD297/DO/DOK1 | |
ART4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
420 | |
DO (Human) Recombinant Protein (Q01) | |
SFDDQYQGCSKQVMEKLTQGDYFTKDIEAQKNYFRMWQKAHLAWLNQGKVLPQNMTTTHAVAILFYTLNSNVHSDFTRAMASVARTPQQYERSFHFKYLH | |
RUO | |
ART4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |