Learn More
Abnova™ Human DKFZP586B1621 Partial ORF (NP_056348, 319 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026007-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined. [provided by RefSeq]
Sequence: VDEPLLKLIDAETTAAAWPNVAAVSITGRKRSRVAPAEPQEAPDSTAAGGSASKRMALVLERVCSTLLGLEEHLNALDRAAGDGDCGTTHSRAARAIQEWLKSpecifications
NP_056348 | |
Liquid | |
26007 | |
DKFZP586B1621 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586B1621/MGC5621 | |
DAK | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VDEPLLKLIDAETTAAAWPNVAAVSITGRKRSRVAPAEPQEAPDSTAAGGSASKRMALVLERVCSTLLGLEEHLNALDRAAGDGDCGTTHSRAARAIQEWLK | |
RUO | |
DAK | |
Wheat Germ (in vitro) | |
GST |