Learn More
Abnova™ Human CUL4B Full-length ORF (AAH36216.1, 1 a.a. - 913 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008450-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the cullin family. The encoded protein forms a complex that functions as an E3 ubiquitin ligase and catalyzes the polyubiquitination of specific protein substrates in the cell. The protein interacts with a ring finger protein, and is required for the proteolysis of several regulators of DNA replication including chromatin licensing and DNA replication factor 1 and cyclin E. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MMSQSSGSGDGNDDEATTSKDGGFSSPSPSAAAAAQEVRSATDGNTSTTPPTSAKKRKLNSSSSSSSNSSNEREDFDSTSSSSSTPPLQPRDSASPSTSSFCLGVSVAASSHVPIQKKLRFEDTLEFVGFDAKMAEESSSSSSSSSPTAATSQQQQLKNKSILISSVASVHHANGLAKSSTTVSSFANSKPGSAKKLVIKNFKDKPKLPENYTDETWQKLKEAVEAIQNSTSIKYNLEELYQAVENLCSYKISANLYKQLRQICEDHIKAQIHQFREDSLDSVLFLKKIDRCWQNHCRQMIMIRSIFLFLDRTYVLQNSMLPSIWDMGLELFRAHIISDQKVQNKTIDGILLLIERERNGEAIDRSLLRSLLSMLSDLQIYQDSFEQRFLEETNRLYAAEGQKLMQEREVPEYLHHVNKRLEEEADRLITYLDQTTQKSLIATVEKQLLGEHLTAILQKGLNNLLDENRIQDLSLLYQLFSRVRGGVQVLLQQWIEYIKAFGSTIVINPEKDKTMVQELLDFKDKVDHIIDICFLKNEKFINAMKEAFETFINKRPNKPAELIAKYVDSKLRAGNKEATDEELEKMLDKIMIIFRFIYGKDVFEAFYKKDLAKRLLVGKSASVDAEKSMLSKLKHECGAAFTSKLEGMFKDMELSKDIMIQFKQYMQNQNVPGNIELTVNILTMGYWPTYVPMEVHLPPEMVKLQEIFKTFYLGKHSGRKLQWQSTIGHCVLKAEFKEGKKELQVSLFQTLVLLMFNEGEEFSLEEIKQATGIEDGELRRTLQSLACGKARVLAKNPKGKDIEDGDKFICNDDFKHKLFRIKINQIQMKETVEEQASTTERVFQDRQYQIDAAIVRIMKMRKTLSHNLLVSEVYNQLKFPVKPADLKKRIESLIDRDYMERDKENPNQYNYIASpecifications
AAH36216.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
130.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F1470/KIAA0695/MRXHF2/MRXSC/SFM2 | |
CUL4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8450 | |
CUL4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMSQSSGSGDGNDDEATTSKDGGFSSPSPSAAAAAQEVRSATDGNTSTTPPTSAKKRKLNSSSSSSSNSSNEREDFDSTSSSSSTPPLQPRDSASPSTSSFCLGVSVAASSHVPIQKKLRFEDTLEFVGFDAKMAEESSSSSSSSSPTAATSQQQQLKNKSILISSVASVHHANGLAKSSTTVSSFANSKPGSAKKLVIKNFKDKPKLPENYTDETWQKLKEAVEAIQNSTSIKYNLEELYQAVENLCSYKISANLYKQLRQICEDHIKAQIHQFREDSLDSVLFLKKIDRCWQNHCRQMIMIRSIFLFLDRTYVLQNSMLPSIWDMGLELFRAHIISDQKVQNKTIDGILLLIERERNGEAIDRSLLRSLLSMLSDLQIYQDSFEQRFLEETNRLYAAEGQKLMQEREVPEYLHHVNKRLEEEADRLITYLDQTTQKSLIATVEKQLLGEHLTAILQKGLNNLLDENRIQDLSLLYQLFSRVRGGVQVLLQQWIEYIKAFGSTIVINPEKDKTMVQELLDFKDKVDHIIDICFLKNEKFINAMKEAFETFINKRPNKPAELIAKYVDSKLRAGNKEATDEELEKMLDKIMIIFRFIYGKDVFEAFYKKDLAKRLLVGKSASVDAEKSMLSKLKHECGAAFTSKLEGMFKDMELSKDIMIQFKQYMQNQNVPGNIELTVNILTMGYWPTYVPMEVHLPPEMVKLQEIFKTFYLGKHSGRKLQWQSTIGHCVLKAEFKEGKKELQVSLFQTLVLLMFNEGEEFSLEEIKQATGIEDGELRRTLQSLACGKARVLAKNPKGKDIEDGDKFICNDDFKHKLFRIKINQIQMKETVEEQASTTERVFQDRQYQIDAAIVRIMKMRKTLSHNLLVSEVYNQLKFPVKPADLKKRIESLIDRDYMERDKENPNQYNYIA | |
RUO | |
CUL4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |