missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CROT Full-length ORF (NP_066974.2, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054677-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120693

  • 359.00€ / 10µg

Estimated Shipment: 21-09-2022
to see stock.
Add to basket

Description

Description

This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4,8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described

Sequence: MENQLAKSTEERTFQYQDSLPSLPVPSLEESLKKYLESVKPFANQEEYKKTEEIVQKFQSGIGEKLHQKLLERAKGKRNWLEEWWLNVAYLDVRIPSQLNVNFAGPAAHFEHYWPPKEGTQLERGSITLWHNLNYWQLLRKEKVPVHKVGNTPLDMNQFRMLFSTCKVPGITRDSIMNYFRTESEGRSPNHIVVLCRGRAFVFDVIHEGCLVTPPELLRQLTYIHKKCHSEPDGPGIAALTSEERTRWAKAREYLIGLDPENLALLEKIQSSLLVYSMEDSSPHVTPEDYSEIIAAILIGDPTVRWGDKSYNLISFSNGVFGCNCDHAPFDAMIMVNISYYVDEKIFQNEGRWKGSEKVRDIPLPEELIFIVDEKVLNDINQAKAQYLREASDLQIAAYAFTSFGKKLTKNKMLHPDTFIQLALQLAYYRLHGHPGCCYETAMTRHFYHGRTETMRSCTVEAVRWCQSMQDPSVNLRERQQKMLQAFAKHNKMMKDCSAGKGFDRHLLGLLLIAKEEGLPVPELFTDPLFSKSGGGGNFVLSTSLVGYLRVQGVVVPMVHNGYGFFYHIRDDRFVVACSAWKSCPETDAEKLVQLTFCAFHDMIQLMNSTHL
Specifications

Specifications

NP_066974.2
Liquid
54677
CROT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MENQLAKSTEERTFQYQDSLPSLPVPSLEESLKKYLESVKPFANQEEYKKTEEIVQKFQSGIGEKLHQKLLERAKGKRNWLEEWWLNVAYLDVRIPSQLNVNFAGPAAHFEHYWPPKEGTQLERGSITLWHNLNYWQLLRKEKVPVHKVGNTPLDMNQFRMLFSTCKVPGITRDSIMNYFRTESEGRSPNHIVVLCRGRAFVFDVIHEGCLVTPPELLRQLTYIHKKCHSEPDGPGIAALTSEERTRWAKAREYLIGLDPENLALLEKIQSSLLVYSMEDSSPHVTPEDYSEIIAAILIGDPTVRWGDKSYNLISFSNGVFGCNCDHAPFDAMIMVNISYYVDEKIFQNEGRWKGSEKVRDIPLPEELIFIVDEKVLNDINQAKAQYLREASDLQIAAYAFTSFGKKLTKNKMLHPDTFIQLALQLAYYRLHGHPGCCYETAMTRHFYHGRTETMRSCTVEAVRWCQSMQDPSVNLRERQQKMLQAFAKHNKMMKDCSAGKGFDRHLLGLLLIAKEEGLPVPELFTDPLFSKSGGGGNFVLSTSLVGYLRVQGVVVPMVHNGYGFFYHIRDDRFVVACSAWKSCPETDAEKLVQLTFCAFHDMIQLMNSTHL
RUO
CROT
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COT
CROT
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers