Learn More
Abnova™ Human COX4I2 Partial ORF (NP_115998, 21 a.a. - 104 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_115998 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84701 |
Molecular Weight (g/mol) | 34.98kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100857
|
Abnova™
H00084701-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16190847
|
Abnova™
H00084701-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes isoform 2 of subunit IV. Isoform 1 of subunit IV is encoded by a different gene, however, the two genes show a similar structural organization. Subunit IV is the largest nuclear encoded subunit which plays a pivotal role in COX regulation. [provided by RefSeq]
Sequence: MHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKTSpecifications
NP_115998 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COX4/COX4-2/COX4B/COX4L2/COXIV-2/dJ857M17.2 | |
COX4I2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84701 | |
COX4I2 (Human) Recombinant Protein (Q01) | |
MHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKT | |
RUO | |
COX4I2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |