missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COQ7 Full-length ORF (NP_057222.2, 1 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_057222.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10229 |
Molecular Weight (g/mol) | 50.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16164682
|
Abnova™
H00010229-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16154682
|
Abnova™
H00010229-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is similar to a mitochondrial di-iron containing hydroxylase in Saccharomyces cerevisiae that is involved with ubiquinone biosynthesis. Mutations in the yeast gene lead to slower development and longer life span. Transcript variants have been described for this gene, but their full-length sequences have not been determined. [provided by RefSeq]
Sequence: MSCAGAAAAPRLWRLRPGARRSLSAYGRRTSVRFRSSGMTLDNISRAAVDRIIRVDHAGEYGANRIYAGQMAVLGRTSVGPVIQKMWDQEKDHLKKFNELMVTFRVRPTVLMPLWNVLGFALGAGTALLGKEGAMACTVAVEESIAHHYNNQIRTLMEEDPEKYEELLQLIKKFRDEELEHHDIGLDHDAELAPAYAVLKSIIQAGCRVAIYLSERLSpecifications
NP_057222.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSCAGAAAAPRLWRLRPGARRSLSAYGRRTSVRFRSSGMTLDNISRAAVDRIIRVDHAGEYGANRIYAGQMAVLGRTSVGPVIQKMWDQEKDHLKKFNELMVTFRVRPTVLMPLWNVLGFALGAGTALLGKEGAMACTVAVEESIAHHYNNQIRTLMEEDPEKYEELLQLIKKFRDEELEHHDIGLDHDAELAPAYAVLKSIIQAGCRVAIYLSERL | |
RUO | |
COQ7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10229 | |
COQ7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAT5/CLK-1/CLK1 | |
COQ7 | |
Recombinant | |
wheat germ expression system |