missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COL16A1 Partial ORF (NP_001847, 117 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001847 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1307 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103401
|
Abnova™
H00001307-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16113401
|
Abnova™
H00001307-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes the alpha chain of type XVI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion. [provided by RefSeq]
Sequence: WYLFQVTDANGYPQISLEVNSQERSLELRAQGQDGDFVSCIFPVPQLFDLRWHKLMLSVAGRVASVHVDCSSASSQPLGPRRPMRPVGHVFLGLDAEQGKSpecifications
NP_001847 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
447AA/FP1572 | |
COL16A1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1307 | |
COL16A1 (Human) Recombinant Protein (Q01) | |
WYLFQVTDANGYPQISLEVNSQERSLELRAQGQDGDFVSCIFPVPQLFDLRWHKLMLSVAGRVASVHVDCSSASSQPLGPRRPMRPVGHVFLGLDAEQGK | |
RUO | |
COL16A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |