missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CNGA3 Full-length ORF (NP_001073347.1, 1 a.a. - 676 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001073347.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1261 |
Molecular Weight (g/mol) | 103.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16163341
|
Abnova™
H00001261-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16153341
|
Abnova™
H00001261-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the cyclic nucleotide-gated cation channel protein family which is required for normal vision and olfactory signal transduction. Mutations in this gene are associated with achromatopsia (rod monochromacy) and color blindness. Two alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: MAKINTQYSHPSRTHLKVKTSDRDLNRAENGLSRAHSSSEETSSVLQPGIAMETRGLADSGQGSFTGQGIARLSRLIFLLRRWAARHVHHQDQGPDSFPDRFRGAELKEVSSQESNAQANVGSQEPADRGRRKKTKKKDAIVVDPSSNLYYRWLTAIALPVFYNWYLLICRACFDELQSEYLMLWLVLDYSADVLYVLDVLVRARTGFLEQGLMVSDTNRLWQHYKTTTQFKLDVLSLVPTDLAYLKVGTNYPEVRFNRLLKFSRLFEFFDRTETRTNYPNMFRIGNLVLYILIIIHWNACIYFAISKFIGFGTDSWVYPNISIPEHGRLSRKYIYSLYWSTLTLTTIGETPPPVKDEEYLFVVVDFLVGVLIFATIVGNVGSMISNMNASRAEFQAKIDSIKQYMQFRKVTKDLETRVIRWFDYLWANKKTVDEKEVLKSLPDKLKAEIAINVHLDTLKKVRIFQDCEAGLLVELVLKLRPTVFSPGDYICKKGDIGKEMYIINEGKLAVVADDGVTQFVVLSDGSYFGEISILNIKGSKSGNRRTANIRSIGYSDLFCLSKDDLMEALTEYPEAKKALEEKGRQILMKDNLIDEELARAGADPKDLEEKVEQLGSSLDTLQTRFARLLAEYNATQMKMKQRLSQLESQVKGGGDKPLADGEVPGDATKTEDKQQSpecifications
NP_001073347.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
103.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAKINTQYSHPSRTHLKVKTSDRDLNRAENGLSRAHSSSEETSSVLQPGIAMETRGLADSGQGSFTGQGIARLSRLIFLLRRWAARHVHHQDQGPDSFPDRFRGAELKEVSSQESNAQANVGSQEPADRGRRKKTKKKDAIVVDPSSNLYYRWLTAIALPVFYNWYLLICRACFDELQSEYLMLWLVLDYSADVLYVLDVLVRARTGFLEQGLMVSDTNRLWQHYKTTTQFKLDVLSLVPTDLAYLKVGTNYPEVRFNRLLKFSRLFEFFDRTETRTNYPNMFRIGNLVLYILIIIHWNACIYFAISKFIGFGTDSWVYPNISIPEHGRLSRKYIYSLYWSTLTLTTIGETPPPVKDEEYLFVVVDFLVGVLIFATIVGNVGSMISNMNASRAEFQAKIDSIKQYMQFRKVTKDLETRVIRWFDYLWANKKTVDEKEVLKSLPDKLKAEIAINVHLDTLKKVRIFQDCEAGLLVELVLKLRPTVFSPGDYICKKGDIGKEMYIINEGKLAVVADDGVTQFVVLSDGSYFGEISILNIKGSKSGNRRTANIRSIGYSDLFCLSKDDLMEALTEYPEAKKALEEKGRQILMKDNLIDEELARAGADPKDLEEKVEQLGSSLDTLQTRFARLLAEYNATQMKMKQRLSQLESQVKGGGDKPLADGEVPGDATKTEDKQQ | |
RUO | |
CNGA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1261 | |
CNGA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACHM2/CCNC1/CCNCa/CCNCalpha/CNCG3/CNG3 | |
CNGA3 | |
Recombinant | |
wheat germ expression system |