missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CMTM1 Full-length ORF (NP_851787.1, 1 a.a. - 114 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_851787.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 113540 |
Molecular Weight (g/mol) | 39kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107013
|
Abnova™
H00113540-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16197003
|
Abnova™
H00113540-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. The protein encoded by this gene may play an important role in testicular development. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MDPEHAKPESSEAPSGNLKQPETAAALSLILGALACFIITQANESFITITSLEICIVVFFILIYVLTLHHLLTYLHWPLLDLTNSIITAVFLSVVAILAMQEKKRRHLLYVGGRSpecifications
NP_851787.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDPEHAKPESSEAPSGNLKQPETAAALSLILGALACFIITQANESFITITSLEICIVVFFILIYVLTLHHLLTYLHWPLLDLTNSIITAVFLSVVAILAMQEKKRRHLLYVGGR | |
RUO | |
CMTM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
113540 | |
CMTM1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKLFH/CKLFH1/CKLFSF1/MGC71870 | |
CMTM1 | |
Recombinant | |
wheat germ expression system |