Learn More
Abnova™ Human CLDN11 Partial ORF (NP_005593, 28 a.a. - 82 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005593 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5010 |
Molecular Weight (g/mol) | 31.79kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112255
|
Abnova™
H00005010-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16102255
|
Abnova™
H00005010-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the claudin family of tight junction associated proteins and is a major component of central nervous system myelin that is necessary for normal CNS function. There is growing evidence that the protein determines the permeability between layers of myelin sheaths via focal adhesion and, with its expression highly regulated during development, may play an important role in cellular proliferation and migration. In addition, the protein is a candidate autoantigen in the development of autoimmune demyelinating disease. [provided by RefSeq]
Sequence: NDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRSpecifications
NP_005593 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.79kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OSP/OTM | |
CLDN11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5010 | |
CLDN11 (Human) Recombinant Protein (Q01) | |
NDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACR | |
RUO | |
CLDN11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |