missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CIRH1A Partial ORF (NP_116219, 587 a.a. - 686 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_116219 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84916 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100947
|
Abnova™
H00084916-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16190937
|
Abnova™
H00084916-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence. [provided by RefSeq]
Sequence: SFHPKRPMHILLHDAYMFCIIDKSLPLPNDKTLLYNPFPPTNESDVIRRRTAHAFKISKIYKPLLFMDLLDERTLVAVERPLDDIIAQLPPPIKKKKFGTSpecifications
NP_116219 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIRHIN/FLJ14728/FLJ17146/KIAA1988/NAIC/TEX292 | |
CIRH1A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84916 | |
CIRH1A (Human) Recombinant Protein (Q01) | |
SFHPKRPMHILLHDAYMFCIIDKSLPLPNDKTLLYNPFPPTNESDVIRRRTAHAFKISKIYKPLLFMDLLDERTLVAVERPLDDIIAQLPPPIKKKKFGT | |
RUO | |
CIRH1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |