missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHODL Partial ORF (NP_079220, 23 a.a. - 94 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_079220 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140578 |
Molecular Weight (g/mol) | 33.66kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101947
|
Abnova™
H00140578-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16191937
|
Abnova™
H00140578-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. [provided by RefSeq]
Sequence: RVVSGQKVCFADFKHPCYKMAYFHELSSRVSFQEARLACESEGGVLLSLENEAEQKLIESMLQNLTKPGTGISpecifications
NP_079220 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf68/FLJ12627/MT75/PRED12 | |
CHODL | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140578 | |
CHODL (Human) Recombinant Protein (Q01) | |
RVVSGQKVCFADFKHPCYKMAYFHELSSRVSFQEARLACESEGGVLLSLENEAEQKLIESMLQNLTKPGTGI | |
RUO | |
CHODL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |