missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CDSN Partial ORF (NP_001255, 306 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001255 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1041 |
Molecular Weight (g/mol) | 31.24kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103091
|
Abnova™
H00001041-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16193081
|
Abnova™
H00001041-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein found in corneodesmosomes, which localize to human epidermis and other cornified squamous epithelia. During maturation of the cornified layers, the protein undergoes a series of cleavages, which are thought to be required for desquamation. The gene is located in the major histocompatibility complex (MHC) class I region on chromosome 6. [provided by RefSeq]
Sequence: YLVPGMTYSKGKIYPVGYFTKENPVKGSPGVPSFAAGPPISEGKYFSSNPSpecifications
NP_001255 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.24kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S586E/HTSS/S | |
CDSN | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1041 | |
CDSN (Human) Recombinant Protein (Q01) | |
YLVPGMTYSKGKIYPVGYFTKENPVKGSPGVPSFAAGPPISEGKYFSSNP | |
RUO | |
CDSN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |