Learn More
ACROBiosystems Human CD38 Protein, Fc Tag
MALS verified
Brand: ACROBiosystems CD8-H5255-50UG
Additional Details : Weight : 0.00001kg
Description
CD antigen CD38 is also known as ADP-ribosyl cyclase 1, which belongs to the ADP-ribosyl cyclase family. CD38 is expressed at high levels in pancreas, liver, kidney, brain, testis, ovary, placenta, malignant lymphoma and neuroblastoma. CD38 is a multifunctional ectoenzyme that catalyzes the synthesis and hydrolysis of cyclic ADP-ribose (cADPR) from NAD+ to ADP-ribose. These reaction products are essential for the regulation of intracellular Ca2+. The loss of CD38 function is associated with impaired immune responses, metabolic disturbances, and behavioral modifications. The CD38 protein is a marker of cell activation. It has been connected to HIV infection, leukemias, myelomas, solid tumors, type II diabetes mellitus and bone metabolism. CD38 has been used as a prognostic marker in leukemia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.5 kDa. The protein migrates as 66-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001766.2 | |
50 μg | |
Val 43 - Ile 300 | |
CD38,T10,cADPr hydrolase 1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
56.5 kDa | |
Human CD38, Fc Tag (CD8-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # P28907-1). | |
-20°C | |
CD38 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |