Learn More
ACROBiosystems Human CD30 / TNFRSF8 Protein, His Tag
MALS verified
Brand: ACROBiosystems CD0-H5229-100UG
Additional Details : Weight : 0.00001kg
Description
Human CD30 is also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. TNFRSF-8 is expressed by activated, but not by resting, T and B cells. Also, CD30 is expressed on classical Hodgkin Lymphoma cells together with CD15. CD30 is the receptor for TNFSF8/CD30L. CD30 can interact with TRAF2 and TRAF5, and mediate the signal transduction that leads to the activation of NF-kappa-B. TNFRSF8 may play a role in the regulation of cellular growth and transformation of activated lymphoblasts. TNFRSF8 is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.3 kDa. The protein migrates as 53-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001234.2 | |
100 µg | |
Phe 19 - Lys 379 | |
TNFRSF8,CD30,D1S166E,Ki-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
39.3 kDa | |
Human CD30, His Tag (CD0-H5229) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). | |
-20°C | |
CD30 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |