Learn More
ACROBiosystems Human CD117 / c-kit Protein, Fc Tag
Brand: ACROBiosystems CD7-H5255-1MG
Additional Details : Weight : 0.00001kg
Description
Mast/stem cell growth factor receptor Kit (c-Kit), a member of the protein kinase superfamily, Tyr protein kinase family and CSF-1/PDGF receptor subfamily, is also known as piebald trait protein (PBT), p145 c-kit, tyrosine-protein kinase Kit and CD117,which contains five Ig-like C2-type (immunoglobulin-like) domains and one protein kinase domain. CD117 acts as cell-surface receptor for the cytokine KITLG/SCF and plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. In response to KITLG/SCF binding, CD117 can activate several signaling pathways. Furthermore, Activated KIT promotes phosphorylation of the protein phosphatases PTPN6/SHP-1 and PTPRU, and of the transcription factors STAT1, STAT3, STAT5A and STAT5B.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. The protein migrates as 100-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
P10721-2 | |
1 mg | |
Gln 26 - Thr 516 | |
CD117,SCFR,c-Kit,KIT | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
81.9 kDa | |
Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2). | |
-20°C | |
CD117 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |