missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCDC28B Full-length ORF (NP_077272.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079140-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104113

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

The product of this gene localizes to centrosomes and basal bodies. It interacts and colocalizes with several proteins associated with Bardet-Biedl syndrome (BBS). [provided by RefSeq]

Sequence: MDDKKKKRSPKPCLAQPAQAPGTLRRVPVPTSHSGSLALGLPHLPSPKQRAKFKRVGKEKCRPVLAGGGSGSAGTPLQHSFLTEVTDVYEMEGGLLNLLNDFHSGRLQAFGKECSFEQLEHVREMQEKLARLHFSLDVCGEEEDDEEEEDGVTEGLPEEQKKTMADRNLDQLLSNLEDLSNSIQKLHLAENAEPEEQSAA
Specifications

Specifications

NP_077272.2
Liquid
79140
CCDC28B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDKKKKRSPKPCLAQPAQAPGTLRRVPVPTSHSGSLALGLPHLPSPKQRAKFKRVGKEKCRPVLAGGGSGSAGTPLQHSFLTEVTDVYEMEGGLLNLLNDFHSGRLQAFGKECSFEQLEHVREMQEKLARLHFSLDVCGEEEDDEEEEDGVTEGLPEEQKKTMADRNLDQLLSNLEDLSNSIQKLHLAENAEPEEQSAA
RUO
CCDC28B
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC1203/MGC16441/RP4-622L5.5
CCDC28B
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers