missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CCDC102B Full-length ORF (NP_079057.1, 1 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_079057.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79839 |
Molecular Weight (g/mol) | 61.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16194533
|
Abnova™
H00079839-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16104543
|
Abnova™
H00079839-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
Sequence: MKPNLDGVDLFNNGGSGNGETKTGLRLKAINLPLENEVTEISALQVHLDEFQKILWKEREMRTALEKEIERLESALSLWKWKYEELKESKPKNVKEFDILLGQHNDEMQELSGNIKEESKSQNSKDRVICELRAELERLQAENTSEWDKREILEREKQGLERENRRLKIQVKEMEELLDKKNRLSANSQSPDFKMSQIDLQEKNQELLNLQHAYYKLNRQYQANIAELTHANNRVDQNEAEVKKLRLRVEELKQGLNQKEDELDDSLNQIRKLQRSLDEEKERNENLETELRHLQNWSpecifications
NP_079057.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKPNLDGVDLFNNGGSGNGETKTGLRLKAINLPLENEVTEISALQVHLDEFQKILWKEREMRTALEKEIERLESALSLWKWKYEELKESKPKNVKEFDILLGQHNDEMQELSGNIKEESKSQNSKDRVICELRAELERLQAENTSEWDKREILEREKQGLERENRRLKIQVKEMEELLDKKNRLSANSQSPDFKMSQIDLQEKNQELLNLQHAYYKLNRQYQANIAELTHANNRVDQNEAEVKKLRLRVEELKQGLNQKEDELDDSLNQIRKLQRSLDEEKERNENLETELRHLQNW | |
RUO | |
CCDC102B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79839 | |
CCDC102B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACY1L/AN/C18orf14/DKFZp434K1426/DKFZp686I08254/FLJ23594/HsT1731/MGC161726/MGC161728 | |
CCDC102B | |
Recombinant | |
wheat germ expression system |