missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CBLB Partial ORF (NP_733762, 21 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000868-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: RILGIIDAIQDAVGPPKQAAADRRTVEKTWKLMDKVVRLCQNPKLQLKNSPPYILDILPDTYQHLRLILSKYDDNQKLAQLSENEYFKIYIDSLMKKSKRAIRLFKEGKSpecifications
NP_733762 | |
Liquid | |
868 | |
CBLB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J10223/DKFZp779A0729/DKFZp779F1443/FLJ36865/FLJ41152/Nbla00127/RNF56 | |
CBLB | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RILGIIDAIQDAVGPPKQAAADRRTVEKTWKLMDKVVRLCQNPKLQLKNSPPYILDILPDTYQHLRLILSKYDDNQKLAQLSENEYFKIYIDSLMKKSKRAIRLFKEGK | |
RUO | |
CBLB | |
Wheat Germ (in vitro) | |
GST |