Learn More
Abnova™ Human CARD14 Partial ORF (NP_077015, 846 a.a. - 945 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_077015 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79092 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16119936
|
Abnova™
H00079092-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16109936
|
Abnova™
H00079092-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: CLLQGFKKCLAEYLSQEEYEAWSQRGDIIQEGEVSGGRCWVTRHAVESLMEKNTHALLDVQLDSVCTLHRMDIFPIVIHVSVNEKMAKKLKKGLQRLGTSSpecifications
NP_077015 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIMP2/CARMA2 | |
CARD14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79092 | |
CARD14 (Human) Recombinant Protein (Q01) | |
CLLQGFKKCLAEYLSQEEYEAWSQRGDIIQEGEVSGGRCWVTRHAVESLMEKNTHALLDVQLDSVCTLHRMDIFPIVIHVSVNEKMAKKLKKGLQRLGTS | |
RUO | |
CARD14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |