missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CAPZA3 Partial ORF (NP_201585, 200 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_201585 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 93661 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16161287
|
Abnova™
H00093661-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16151287
|
Abnova™
H00093661-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes an actin capping protein, one of the F-actin capping protein alpha subunit family. The encoded protein is predominantly localized to the neck region of ejaculated sperm, other immunohistochemical signals were found in the tail and postacrosomal regions. The encoded protein may also form heterodimers of alpha and beta subunits. This protein may be important in determining sperm architecture and male fertility. [provided by RefSeq]
Sequence: NLHIEISKDLKESLEIVNQAQLALSFARLVEEQENKFQAAVLEELQELSNEALRKILRRDLPVTRTLIDWHRILSDLNLVMYPKLGYVIYSRSVLCNWIISpecifications
NP_201585 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPPA3/Gsg3 | |
CAPZA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
93661 | |
CAPZA3 (Human) Recombinant Protein (Q01) | |
NLHIEISKDLKESLEIVNQAQLALSFARLVEEQENKFQAAVLEELQELSNEALRKILRRDLPVTRTLIDWHRILSDLNLVMYPKLGYVIYSRSVLCNWII | |
RUO | |
CAPZA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |